Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Triisopropyl borate ≥98%

Supplier: BEANTOWN CHEMICAL - BTC

142780-100ML 142780-2.5L 142780-500ML
BT142780-100MLEA 49 USD InStock
BT142780-100ML BT142780-2.5L BT142780-500ML
Triisopropyl borate ≥98%
Triisopropyl borate
CAS: 5419-55-6; EC No: 226-529-9; MDL No: MFCD00008872; RTECS: ED5950000 UN No: UN2616; Haz Class: 3; Packing Group: II Liquid; Linear Formula: [(CH3)2CHO]3B; Molecular Formula: C9H21BO3; MW: 188.08 Melting Point: -59°; Boiling Point: 139-141°; Flash point: 17°C (63°F) Density (g/mL): 0.815; Refractive Index: 1.376 Moisture Sensitive

  • RTECS: ED5950000
  • Refractive index: 1.376
  • Moisture Sensitive
  • Liquid


Formula: C₉H₂₁BO₃
MW: 188.07 g/mol
Boiling Pt: 139
Melting Pt: -59
Density: 0.815 g/mL
Flash Pt: 17 °C (63 °F)
MDL Number: MFCD00008872
CAS Number: 5419-55-6
EINECS: 226-529-9
UN: 2616
ADR: 3 II
Order Now
Specification Test Results
Assay 98%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty