Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Indole ≥99%

Supplier: BEANTOWN CHEMICAL - BTC

142845-1KG 142845-250G 142845-50G
BT142845-1KGEA 429 USD InStock
BT142845-1KG BT142845-250G BT142845-50G
Indole ≥99%
Indole
CAS: 120-72-9; EC No: 204-420-7; MDL No: MFCD00005607; RTECS: NL2450000 UN No: UN2811; Haz Class: 6.1; Packing Group: III Solid; Molecular Formula: C8H7N; MW: 117.15 Melting Point: 51-54°; Boiling Point: 253-254°; Flash point: 121°C (249°F) Light Sensitive

  • RTECS: NL2450000
  • Light Sensitive
  • Solid


Formula: C₈H₇N
MW: 117.15 g/mol
Boiling Pt: 253…254 °C
Melting Pt: 51…54 °C
Flash Pt: 121 °C (249 °F)
MDL Number: MFCD00005607
CAS Number: 120-72-9
EINECS: 204-420-7
UN: 2811
ADR: 6.1 III
Merck Index: 13,04985
Order Now
Specification Test Results
Assay 99%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty