Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Copper ≥99.5% (trace metals basis), spherical powder -100 mesh

Supplier: BEANTOWN CHEMICAL - BTC

143185-1KG 143185-250G 143185-50G
BT143185-1KGEA 98 USD InStock
BT143185-1KG BT143185-250G BT143185-50G
Copper ≥99.5% (trace metals basis), spherical powder -100 mesh
Copper
CAS: 7440-50-8; EC No: 231-159-6; MDL No: MFCD00010965 UN No: UN3089; Haz Class: 4.1; Packing Group: III Powder; Molecular Formula: Cu; MW: 63.55 Melting Point: 1083.4°; Boiling Point: 2567° Density (g/mL): 8.94

  • Powder


Formula: Cu
MW: 63.55 g/mol
Boiling Pt: 2595
Melting Pt: 1083
Density: 8.94 g/mL
MDL Number: MFCD00010965
CAS Number: 7440-50-8
EINECS: 231-159-6
UN: 3089
ADR: 4.1 III
Merck Index: 13,02545
Order Now
Specification Test Results
Assay 99.5%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty