Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N,N-Dimethylformamide ≥99%

Supplier: BEANTOWN CHEMICAL - BTC

143245-10L 143245-2.5L 143245-250ML
BT143245-10LEA 306.21 USD InStock
BT143245-10L BT143245-2.5L BT143245-250ML
N,N-Dimethylformamide ≥99%
N,N-Dimethylformamide
CAS: 68-12-2; EC No: 200-679-5; MDL No: MFCD00003284; RTECS: LQ2100000 UN No: UN2265; Haz Class: 3; Packing Group: III Liquid; Linear Formula: HCON(CH3)2 ; Molecular Formula: C3H7NO; MW: 73.09 Melting Point: -61°; Boiling Point: 153°; Flash point: 57°C (135°F) Density (g/mL): 0.944; Refractive Index: 1.430

  • RTECS: LQ2100000
  • Refractive index: 1.430
  • Liquid


Formula: HCON(CH₃)₂
MW: 73.09 g/mol
Boiling Pt: 153
Melting Pt: -61
Density: 0.944 g/mL
Flash Pt: 57 °C (135 °F)
MDL Number: MFCD00003284
CAS Number: 68-12-2
EINECS: 200-679-5
UN: 2265
ADR: 3 III
Merck Index: 13,03269
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1ac5c5e9758b6c40ccb0cf127b6a6e1c6d608c64&vid=fb9c58451426a2acf92f6525238f8198a167f088&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Assay 99%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty