Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Decyne ≥98%

Supplier: BEANTOWN CHEMICAL - BTC

143740-100G 143740-25G 143740-5G
BT143740-100GEA 314 USD InStock
BT143740-100G BT143740-25G BT143740-5G
1-Decyne ≥98%
1-Decyne
CAS: 764-93-2; EC No: 212-132-8; MDL No: MFCD00009576 UN No: UN3295; Haz Class: 3; Packing Group: III Liquid; Linear Formula: HC≡C(CH2)7CH3; Molecular Formula: C10H18; MW: 138.25 Melting Point: -44°; Boiling Point: 174°; Flash point: 48°C (118°F) Density (g/mL): 0.766; Refractive Index: 1.427

  • Refractive index: 1.427
  • Liquid


Formula: C₁₀H₁₈
MW: 138.25 g/mol
Boiling Pt: 174
Melting Pt: -44
Density: 0.766 g/mL
Flash Pt: 48 °C (118 °F)
MDL Number: MFCD00009576
CAS Number: 764-93-2
EINECS: 212-132-8
UN: 3295
ADR: 3 III
Order Now
Specification Test Results
Assay 98%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty