144150-1G
144150-5G
BT144150-1GEA
57.08
USD
InStock
BT144150-1G
BT144150-5G
Tin(II) trifluoromethanesulphonate ≥97%
Tin(II) trifluoromethanesulphonate
CAS: 62086-04-8; MDL No: MFCD00191251
UN No: UN3288; Haz Class: 6.1; Packing Group: III
Powder; Linear Formula: (CF3SO3)2Sn; Molecular Formula: C2F6O6S2Sn; MW: 416.85
Melting Point: 300°
Moisture Sensitive
- Moisture Sensitive
- Powder