Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(±)-Ethyl-2-bromobutyrate ≥97%

Supplier: BEANTOWN CHEMICAL - BTC

144340-100G 144340-250G
BT144340-100GEA 38 USD InStock
BT144340-100G BT144340-250G
(±)-Ethyl-2-bromobutyrate ≥97%
(±)-Ethyl-2-bromobutyrate
CAS: 533-68-6; EC No: 208-574-6; MDL No: MFCD00000155 UN No: UN3272; Haz Class: 3; Packing Group: III Liquid; Linear Formula: CH3CH2CHBrCOOCH2CH3; Molecular Formula: C6H11BrO2 ; MW: 195.05 Boiling Point: 177°; Flash point: 56°C (133°F) Density (g/mL): 1.321; Refractive Index: 1.447 Light Sensitive

  • Refractive index: 1.447
  • Light Sensitive
  • Liquid


Formula: C₆H₁₁BrO₂
Boiling Pt: 177 °C
Density: 1.321 g/mL
Flash Pt: 56 °C (133 °F)
MDL Number: MFCD00000155
CAS Number: 533-68-6
EINECS: 208-574-6
UN: 3272
ADR: 3,III
Order Now
Specification Test Results
Assay 97%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty