Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Allyl cyanide ≥90%, Technical Grade

Supplier: BEANTOWN CHEMICAL - BTC

144395-100G 144395-25G
BT144395-100GEA 142 USD InStock
BT144395-100G BT144395-25G
Allyl cyanide ≥90%, Technical Grade
Allyl cyanide
CAS: 109-75-1; EC No: 203-701-1; MDL No: MFCD00001962; RTECS: EM8050000 UN No: UN1992; Haz Class: 3 (6.1); Packing Group: III Liquid; Linear Formula: CH2=CHCH2CN; Molecular Formula: C4H5N; MW: 67.09 Boiling Point: 116-121°; Flash point: 27°C (75°F) Density (g/mL): 0.834; Refractive Index: 1.405

  • RTECS: EM8050000
  • Refractive index: 1.405
  • Liquid


Formula: C₄H₅N
MW: 67.09 g/mol
Boiling Pt: 116…121 °C
Density: 0.834 g/mL
Flash Pt: 27 °C (75 °F)
MDL Number: MFCD00001962
CAS Number: 109-75-1
EINECS: 203-701-1
UN: 1992
ADR: 3 III
Order Now
Specification Test Results
Assay 90%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty