Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Titanium(IV) fluoride 98

Supplier: BEANTOWN CHEMICAL - BTC

144875-100G 144875-25G
BT144875-100GEA 314 USD InStock
BT144875-100G BT144875-25G
Titanium(IV) fluoride 98
Titanium(IV) fluoride
CAS: 7783-63-3; EC No: 232-017-6; MDL No: MFCD00042545 UN No: UN3260; Haz Class: 8; Packing Group: II Crystalline; Linear Formula: TiF4; MW: 123.87 Melting Point: 284° (sublimes) Density (g/mL): 2.798 Moisture Sensitive

  • Moisture Sensitive
  • Crystalline


Formula: F₄Ti
MW: 123.87 g/mol
Boiling Pt: 284
Melting Pt: 400 (subl.)
Density: 2.798 g/mL
MDL Number: MFCD00042545
CAS Number: 7783-63-3
EINECS: 232-017-6
UN: 3260
ADR: 8 II
Merck Index: 12,09619
Order Now
Specification Test Results
Assay 98%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty