Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,4-Dihydro-2H-pyran ≥99%

Supplier: BEANTOWN CHEMICAL - BTC

145390-100ML 145390-25ML 145390-500ML
BT145390-100MLEA 34.1 USD InStock
BT145390-100ML BT145390-25ML BT145390-500ML
3,4-Dihydro-2H-pyran ≥99%
3,4-Dihydro-2H-pyran
CAS: 110-87-2; EC No: 203-810-4; MDL No: MFCD00006558 UN No: UN2376; Haz Class: 3; Packing Group: II Liquid; Molecular Formula: C5H8O; MW: 84.12 Melting Point: -70°; Boiling Point: 86°; Flash point: -16°C (3°F) Density (g/mL): 0.922; Refractive Index: 1.440

  • Refractive index: 1.440
  • Liquid


Formula: C₅H₈O
MW: 84.12 g/mol
Boiling Pt: 86 °C
Melting Pt: –70 °C
Density: 0.922 g/mL
Flash Pt: –16 °C (3 °F)
MDL Number: MFCD00006558
CAS Number: 110-87-2
EINECS: 203-810-4
UN: 2376
ADR: 3 II
Order Now
Specification Test Results
Assay 99%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty