Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Lauroyl chloride ≥98%

Supplier: BEANTOWN CHEMICAL - BTC

145910-100G 145910-500G
BT145910-100GEA 48.54 USD InStock
BT145910-100G BT145910-500G
Lauroyl chloride ≥98%
Lauroyl chloride
CAS: 112-16-3; EC No: 203-941-7; MDL No: MFCD00000740 UN No: UN3265; Haz Class: 8; Packing Group: II Liquid; Linear Formula: CH3(CH2)10COCl; Molecular Formula: C12H23ClO; MW: 218.77 Boiling Point: 134-137°/11 mmHg; Flash point: 140°C (284°F) Density (g/mL): 0.946; Refractive Index: 1.445 Moisture Sensitive

  • Refractive index: 1.445
  • Moisture Sensitive
  • Liquid


Formula: C₁₂H₂₃ClO
MW: 218.77 g/mol
Boiling Pt: 267
Melting Pt: -12
Density: 0.946 g/mL
Flash Pt: 140 °C (284 °F)
MDL Number: MFCD00000740
CAS Number: 112-16-3
EINECS: 203-941-7
UN: 3265
ADR: 8 II
Order Now
Specification Test Results
Assay 98%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty