Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Copper(I) bromide ≥98%

Supplier: BEANTOWN CHEMICAL - BTC

211040-1KG 211040-250G
BT211040-1KGEA 142 USD InStock
BT211040-1KG BT211040-250G
Copper(I) bromide ≥98%
Copper(I) bromide
CAS: 7787-70-4; EC No: 232-131-6; MDL No: MFCD00010969 UN No: UN3077; Haz Class: 9; Packing Group: III Powder; Linear Formula: CuBr; MW: 143.45 Melting Point: 504° Density (g/mL): 4.71 Air Sensitive

  • Air Sensitive
  • Powder


Formula: BrCu
MW: 143.45
Boiling Pt: 1345
Melting Pt: 504
Density: 4.71 g/mL
MDL Number: MFCD00010969
CAS Number: 7787-70-4
EINECS: 232-131-6
ADR: 9 III
Merck Index: 13,02689
Order Now
Specification Test Results
Assay 98%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty