211055-100ML
211055-2.5L
211055-500ML
BT211055-100MLEA
34.18
USD
InStock
BT211055-100ML
BT211055-2.5L
BT211055-500ML
Cyclohexylamine ≥98%
Cyclohexylamine
CAS: 108-91-8; EC No: 203-629-0; MDL No: MFCD00001486; RTECS: GX0700000
UN No: UN2357; Haz Class: 8 (3); Packing Group: II
Liquid; Molecular Formula: C6H13N; MW: 99.18
Melting Point: -17°; Boiling Point: 134°; Flash point: 27°C (81°F)
Density (g/mL): 0.867; Refractive Index: 1.459
Air Sensitive
- RTECS: GX0700000
- Refractive index: 1.459
- Air Sensitive
- Liquid