Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,4-Epoxy-1-butene ≥98%

Supplier: BEANTOWN CHEMICAL - BTC

211525-1G 211525-25G 211525-5G
BT211525-1GEA 46 USD InStock
BT211525-1G BT211525-25G BT211525-5G
3,4-Epoxy-1-butene ≥98%
3,4-Epoxy-1-butene
CAS: 930-22-3; EC No: 213-210-4; MDL No: MFCD00005149; RTECS: EM7350000 UN No: UN1993; Haz Class: 3; Packing Group: II Liquid; Molecular Formula: C4H6O; MW: 70.09 Boiling Point: 65-66°; Flash point: -50°C (-58°F) Density (g/mL): 0.87; Refractive Index: 1.417 Moisture Sensitive

  • RTECS: EM7350000
  • Refractive index: 1.417
  • Moisture Sensitive
  • Liquid


Formula: C₄H₆O
Boiling Pt: 65…66 °C
Density: 0.87 g/mL
Flash Pt: –50 °C (–58 °F)
MDL Number: MFCD00005149
CAS Number: 930-22-3
EINECS: 213-210-4
UN: 1993
ADR: 3,II
Order Now
Specification Test Results
Assay 98%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty