Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Sodium iodate ≥99%

Supplier: BEANTOWN CHEMICAL - BTC

212330-100G 212330-500G
BT212330-100GEA 50.38 USD InStock
BT212330-100G BT212330-500G
Sodium iodate ≥99%
Sodium iodate
CAS: 7681-55-2; EC No: 231-672-5; MDL No: MFCD00003533 UN No: UN1479; Haz Class: 5.1; Packing Group: II Powder; Linear Formula: NaIO3; MW: 197.89 Density (g/mL): 4.28 Air Sensitive, Light Sensitive, Moisture Sensitive

  • Air Sensitive, Light Sensitive, Moisture Sensitive
  • Powder


Formula: INaO₃
Density: 4.28 g/mL
MDL Number: MFCD00003533
CAS Number: 7681-55-2
EINECS: 231-672-5
UN: 1479
ADR: 5.1,II
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=806173453dcec1d2f791367d269d662d3459149a&vid=806173453dcec1d2f791367d269d662d3459149a&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Assay 99%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty