Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Hexachloroplatinic(IV) acid (20% Pt) (w/w) in water

Supplier: BEANTOWN CHEMICAL - BTC

212400-C2G 212400-C500MG
BT212400-C2GEA 643 USD InStock
BT212400-C2G BT212400-C500
Hexachloroplatinic(IV) acid (20% Pt) (w/w) in water
Hexachloroplatinic(IV) acid
CAS: 16941-12-1; EC No: 241-010-7; MDL No: MFCD00011330 UN No: UN3264; Haz Class: 8; Packing Group: III Liquid; Linear Formula: H2PtCl6; MW: 409.82

  • Liquid


Formula: H₂Cl₆Pt
MDL Number: MFCD00011330
CAS Number: 16941-12-1
EINECS: 241-010-7
UN: 3264
ADR: 8,III
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty