Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Barium hydroxide monohydrate ≥95%

Supplier: BEANTOWN CHEMICAL - BTC

212425-100G 212425-500G
BT212425-100GEA 26 USD InStock
BT212425-100G BT212425-500G
Barium hydroxide monohydrate ≥95%
Barium hydroxide monohydrate
CAS: 22326-55-2; EC No: 241-234-5; MDL No: MFCD00149151; RTECS: CQ9200000 UN No: UN3262; Haz Class: 8; Packing Group: III Crystalline; Linear Formula: Ba(OH)2·H2O; MW: 189.36 Density (g/mL): 3.743 Air Sensitive , Hygroscopic

  • RTECS: CQ9200000
  • Air Sensitive, Hygroscopic
  • Crystalline


Formula: H₂BaO₂·1H₂O
MW: 189.36
Density: 3.743 g/mL
MDL Number: MFCD00149151
CAS Number: 22326-55-2
EINECS: 241-234-5
UN: 3262
ADR: 8 III
Order Now
Specification Test Results
Assay 95%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty