Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Hydrochloric acid 4 M in 1,4-dioxane ≥99%

Supplier: BEANTOWN CHEMICAL - BTC

212670-100ML 212670-50ML
BT212670-100MLEA 109.47 USD InStock
BT212670-100ML BT212670-50ML
Hydrochloric acid 4 M in 1,4-dioxane ≥99%
Hydrochloric acid
CAS: 7647-01-0; EC No: 231-595-7; MDL No: MFCD00011324 UN No: UN2924; Haz Class: 3(8); Packing Group: II Liquid; Molecular Formula: HCl; MW: 36.46 Flash point: 17°C (63°F) Density (g/mL): 1.05 Hygroscopic

  • Hygroscopic
  • Liquid


Formula: HCl
Density: 1.05 g/mL
Flash Pt: 17 °C (63 °F)
MDL Number: MFCD00011324
CAS Number: 7647-01-0
EINECS: 231-595-7
UN: 2924
ADR: 8,II
Order Now
Specification Test Results
Assay 99%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty