Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Germanium(IV) methoxide ≥98%

Supplier: BEANTOWN CHEMICAL - BTC

212755-25G 212755-5G
BT212755-25GEA 858 USD InStock
BT212755-25G BT212755-5G
Germanium(IV) methoxide ≥98%
Germanium(IV) methoxide
CAS: 992-91-6; MDL No: MFCD00014881 UN No: UN2929; Haz Class: 6.1 (3); Packing Group: II Liquid; Linear Formula: Ge(OCH3)4; MW: 196.78 Melting Point: -18°; Boiling Point: 66-67°/36 mmHg; Flash point: 41°C (106°F) Density (g/mL): 1.3254; Refractive Index: 1.399 Moisture Sensitive

  • Refractive index: 1.399
  • Moisture Sensitive
  • Liquid


Formula: C₄H₁₂GeO₄
MW: 196.75
Boiling Pt: 66…67 °C/36 mmHg
Melting Pt: –18 °C
Density: 1.3254 g/mL
Flash Pt: 41 °C (106 °F)
MDL Number: MFCD00014881
CAS Number: 992-91-6
UN: 2929
ADR: 6.1 II
Order Now
Specification Test Results
Assay 98%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty