Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Manganese ≥99.95% (trace metals basis), powder -325 mesh

Supplier: BEANTOWN CHEMICAL - BTC

212840-100G 212840-25G
BT212840-100GEA 190.95 USD InStock
BT212840-100G BT212840-25G
Manganese ≥99.95% (trace metals basis), powder -325 mesh
Manganese
CAS: 7439-96-5; EC No: 231-105-1; MDL No: MFCD00011111; RTECS: OO9625000 UN No: UN3089; Haz Class: 4.1; Packing Group: III Powder; Molecular Formula: Mn; MW: 54.94 Melting Point: 1244°; Boiling Point: 1962° Density (g/mL): 7.3

  • RTECS: OO9625000
  • Powder


Formula: Mn
MW: 54.94 g/mol
Boiling Pt: 1962 °C
Melting Pt: 1244 °C
Density: 7.3 g/mL
MDL Number: MFCD00011111
CAS Number: 7439-96-5
EINECS: 231-105-1
UN: 3089
ADR: 4.3 III
Order Now
Specification Test Results
Assay 99.95%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty