Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4,6-Dimethylpyrimidine ≥94%

Supplier: BEANTOWN CHEMICAL - BTC

213045-1G 213045-5G
BT213045-1GEA 43 USD InStock
BT213045-1G BT213045-5G
4,6-Dimethylpyrimidine ≥94%
4,6-Dimethylpyrimidine
CAS: 1558-17-4; EC No: 216-314-8; MDL No: MFCD00006116 UN No: UN1993; Haz Class: 3; Packing Group: III Liquid; Molecular Formula: C6H8N2; MW: 108.14 Boiling Point: 154°; Flash point: 46°C (114°F) Density (g/mL): 0.98; Refractive Index: 1.490

  • Refractive index: 1.490
  • Liquid


Formula: C₆H₈N₂
Boiling Pt: 154 °C
Density: 0.98 g/mL
Flash Pt: 46 °C (114 °F)
MDL Number: MFCD00006116
CAS Number: 1558-17-4
EINECS: 216-314-8
ADR: 3,III
Order Now
Specification Test Results
Assay 94%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty