213150-1G
213150-5G
BT213150-1GEA
133
USD
InStock
BT213150-1G
BT213150-5G
2-Pyrazinecarbonyl chloride ≥90%, Technical Grade
2-Pyrazinecarbonyl chloride
CAS: 19847-10-0; EC No: 243-367-4; MDL No: MFCD00792886
UN No: UN3261; Haz Class: 8; Packing Group: II
Powder; Molecular Formula: C5H3ClN2O; MW: 142.55
Moisture Sensitive
- Moisture Sensitive
- Powder