213235-1G
213235-25G
213235-5G
BT213235-1GEA
32
USD
InStock
BT213235-1G
BT213235-25G
BT213235-5G
3-(Trimethylsilyl)-2-propyn-1-ol ≥98%
3-(Trimethylsilyl)-2-propyn-1-ol
CAS: 5272-36-6; EC No: 226-094-5; MDL No: MFCD00002913
UN No: UN2810; Haz Class: 6.1; Packing Group: III
Liquid; Molecular Formula: C6H12OSi; MW: 128.24
Boiling Point: 76°/11 mmHg; Flash point: 67°C (153°F)
Density (g/mL): 0.865; Refractive Index: 1.451
Moisture Sensitive
- Refractive index: 1.451
- Moisture Sensitive
- Liquid