Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Nitrophenyl chloroformate ≥97%

Supplier: BEANTOWN CHEMICAL - BTC

213255-100G 213255-25G 213255-5G
BT213255-100GEA 279 USD InStock
BT213255-100G BT213255-25G BT213255-5G
4-Nitrophenyl chloroformate ≥97%
4-Nitrophenyl chloroformate
CAS: 7693-46-1; EC No: 231-706-9; MDL No: MFCD00007321 UN No: UN3261; Haz Class: 8; Packing Group: II Powder; Molecular Formula: C7H4ClNO4; MW: 201.56 Melting Point: 77-79°; Boiling Point: 159-162°/19 mmHg Moisture Sensitive

  • Moisture Sensitive
  • Powder


Formula: C₇H₄ClNO₄
MW: 201.57 g/mol
Boiling Pt: 159
Melting Pt: 77…79 °C
MDL Number: MFCD00007321
CAS Number: 7693-46-1
EINECS: 231-706-9
UN: 3261
ADR: 8 II
Order Now
Specification Test Results
Assay 97%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty