Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Propagyl amine hydrochloride ≥95%

Supplier: BEANTOWN CHEMICAL - BTC

213490-1G 213490-5G
BT213490-1GEA 37 USD InStock
BT213490-1G BT213490-5G
Propagyl amine hydrochloride ≥95%
Propagyl amine hydrochloride
CAS: 15430-52-1; EC No: 239-440-5; MDL No: MFCD00012907 UN No: UN2811; Haz Class: 6.1; Packing Group: II Powder; Linear Formula: HC≡CCH2NH2·HCl; Molecular Formula: C3H5N·HCl; MW: 91.54 Melting Point: 179-182° Hygroscopic

  • Hygroscopic
  • Powder


Formula: C₃H₆ClN
Melting Pt: 179…182 °C
MDL Number: MFCD00012907
CAS Number: 15430-52-1
EINECS: 239-440-5
UN: 2811
ADR: 6.1,II
Order Now
Specification Test Results
Assay 95%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty