Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Yttrium (III) nitrate hydrate ≥99.99%

Supplier: BEANTOWN CHEMICAL - BTC

213830-25G 213830-5G
BT213830-25GEA 110 USD InStock
BT213830-25G BT213830-5G
Yttrium (III) nitrate hydrate ≥99.99%
Yttrium (III) nitrate hydrate


Formula: N₃O₉Y·H₂O
MW: 274.92 g/mol (anhy)
MDL Number: MFCD00011469
CAS Number: 140191-40-8
UN: 1477
ADR: 5.1 III
Order Now
Specification Test Results
Assay 99.99%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty