Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Aluminium nitrate hydrate 99,999

Supplier: BEANTOWN CHEMICAL - BTC

214055-100G 214055-25G 214055-5G
BT214055-100GEA 236.27 USD InStock
BT214055-100G BT214055-25G BT214055-5G
Aluminium nitrate hydrate 99,999
Aluminium nitrate hydrate
CAS: 25838-59-9; EC No: 236-751-8; MDL No: MFCD00003419 UN No: UN1438; Haz Class: 5.1; Packing Group: III Crystalline; Linear Formula: Al(NO3)3·xH2O ; MW: 212.99 (anhydrous) Melting Point: 73° Hygroscopic

  • Hygroscopic
  • Crystalline


Formula: AlN₃O₉·H₂O
MW: 231.01 g/mol
Melting Pt: 73 °C
MDL Number: MFCD00003419
CAS Number: 25838-59-9
EINECS: 236-751-8
UN: 1438
ADR: 5.1 III
Order Now
Specification Test Results
Assay 99.999%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty