215040-100G
215040-25G
215040-5G
BT215040-100GEA
851
USD
InStock
BT215040-100G
BT215040-25G
BT215040-5G
Copper(II) acetate ≥99.999%
Copper(II) acetate
CAS: 142-71-2; EC No: 205-553-3; MDL No: MFCD00008690; RTECS: AG3480000
UN No: UN3077; Haz Class: 9; Packing Group: III
Powder; Linear Formula: Cu(OOCCH3)2; MW: 181.63
Hygroscopic
- RTECS: AG3480000
- Hygroscopic
- Powder