Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Thallium(III) nitrate trihydrate ≥99.5%

Supplier: BEANTOWN CHEMICAL - BTC

215090-25G 215090-5G
BT215090-25GEA 301 USD InStock
BT215090-25G BT215090-5G
Thallium(III) nitrate trihydrate ≥99.5%
Thallium(III) nitrate trihydrate
CAS: 13453-38-8; EC No: 237-325-4; MDL No: MFCD00149874; RTECS: QV2000000 UN No: UN2727; Haz Class: 6.1; Packing Group: II Crystalline; Linear Formula: Tl(NO3)3·3H2O; MW: 444.43 Melting Point: 102-105° (decomposes) Hygroscopic

  • RTECS: QV2000000
  • Hygroscopic
  • Crystalline


Formula: N₃O₉Tl·3H₂O
MW: 444.44
Melting Pt: 102…105 °C (dec.)
MDL Number: MFCD00149874
CAS Number: 13453-38-8
EINECS: 237-325-4
UN: 2727
ADR: 6.1 II
Order Now
Specification Test Results
Assay 99.5%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty