Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Dimethyl diselenide ≥99%

Supplier: BEANTOWN CHEMICAL - BTC

216230-1G 216230-5G
BT216230-1GEA 63 USD InStock
BT216230-1G BT216230-5G
Dimethyl diselenide ≥99%
Dimethyl diselenide
CAS: 7101-31-7; EC No: 230-410-7; MDL No: MFCD00008254 UN No: UN2929; Haz Class: 6.1 (3); Packing Group: II Liquid; Linear Formula: (CH3)2Se2 ; MW: 187.99 Boiling Point: 155-157°; Flash point: 60°C (140°F) Density (g/mL): 1.987 ; Refractive Index: 1.639 Air Sensitive, Moisture Sensitive

  • Refractive index: 1.639
  • Air Sensitive, Moisture Sensitive
  • Liquid


Formula: C₂H₆Se₂
MW: 187.99
Boiling Pt: 155
Melting Pt: -82
Density: 1.987 g/mL
Flash Pt: 60 °C (140 °F)
MDL Number: MFCD00008254
CAS Number: 7101-31-7
EINECS: 230-410-7
UN: 2929
ADR: 6.1 II
Order Now
Specification Test Results
Assay 99%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty