Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Trimethyloxonium tetrafluoroborate ≥96%

Supplier: BEANTOWN CHEMICAL - BTC

216255-10G 216255-250G 216255-50G
BT216255-10GEA 78.02 USD InStock
BT216255-10G BT216255-250G BT216255-50G
Trimethyloxonium tetrafluoroborate ≥96%
Trimethyloxonium tetrafluoroborate
CAS: 420-37-1; EC No: 206-994-4; MDL No: MFCD00011798 UN No: UN3261; Haz Class: 8; Packing Group: II Solid; Linear Formula: O(CH3)3BF4; Molecular Formula: C3H9BF4O; MW: 147.91 Melting Point: 190° (decomposes) Moisture Sensitive

  • Moisture Sensitive
  • Solid


Formula: C₃H₉BF₄O
Melting Pt: 190 °C (dec.)
MDL Number: MFCD00011798
CAS Number: 420-37-1
EINECS: 206-994-4
UN: 3261
ADR: 8,II
Order Now
Specification Test Results
Assay 96%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty