Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Acetonitrile ≥99.5% ACS

Supplier: BEANTOWN CHEMICAL - BTC

216530-1L 216530-4L 216530-500ML
BT216530-1LEA 67.42 USD InStock
BT216530-1L BT216530-4L BT216530-500ML
Acetonitrile ≥99.5% ACS
Acetonitrile
CAS: 75-05-8; EC No: 200-835-2; MDL No: MFCD00001878; RTECS: AL7700000 UN No: UN1648; Haz Class: 3; Packing Group: II Liquid; Linear Formula: CH3CN; MW: 41.05 Melting Point: -48°; Boiling Point: 81-82°; Flash point: 2°C (36°F) Density (g/mL): 0.786; Refractive Index: 1.344

  • RTECS: AL7700000
  • Refractive index: 1.344
  • Liquid


Formula: H₃CCN
MW: 41.05 g/mol
Boiling Pt: 81.6
Melting Pt: -45.7
Density: 0.786 g/mL
Flash Pt: 2 °C (36 °F)
MDL Number: MFCD00001878
CAS Number: 75-05-8
EINECS: 200-835-2
UN: 1648
ADR: 3 II
Merck Index: 13,00071
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cef8be50fcca82050c5bda827ad9c5f57f4c5e27&vid=b965ef40b6e61dd29b33363d25c3df2038fea764&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Assay 99.5% min.
Color (APHA) ≤10
Residue after evaporation ≤0.005%
Titrable acid ≤0.008meq/g
Titrable base ≤0.0006meq/g
Water ≤0.3%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty