216590-100G
216590-25G
216590-500G
BT216590-100GEA
292
USD
InStock
BT216590-100G
BT216590-25G
BT216590-500G
Copper(II) oxide ≥99.995%
Copper(II) oxide
CAS: 1317-38-0; EC No: 215-269-1; MDL No: MFCD00010979; RTECS: GL7900000
UN No: UN3077; Haz Class: 9; Packing Group: III
Powder; Molecular Formula: CuO; MW: 79.55
Melting Point: 1330°
Density (g/mL): 6.4