Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Zirconyl nitrate hydrate ≥99.9%

Supplier: BEANTOWN CHEMICAL - BTC

217385-100G 217385-25G 217385-500G
BT217385-100GEA 207.71 USD InStock
BT217385-100G BT217385-25G BT217385-500G
Zirconyl nitrate hydrate ≥99.9%
Zirconyl nitrate hydrate
CAS: 14985-18-3; EC No: 237-529-3; MDL No: MFCD00149896 UN No: UN1477; Haz Class: 5.1; Packing Group: III Crystalline; Linear Formula: ZrO(NO3)2·xH2O; MW: 231.23 (anhydrous)

  • Crystalline


Formula: N₂O₇Zr·H₂O
MDL Number: MFCD00149896
CAS Number: 14985-18-3
EINECS: 237-529-3
UN: 1477
ADR: 5.1,III
Order Now
Specification Test Results
Assay 99.9%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty