Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Sodium borohydride ≥98%

Supplier: BEANTOWN CHEMICAL - BTC

217465-100G 217465-500G
BT217465-100GEA 91.62 USD InStock
BT217465-100G BT217465-500G
Sodium borohydride ≥98%
Sodium borohydride
CAS: 16940-66-2; EC No: 241-004-4; MDL No: MFCD00003518; RTECS: ED3325000 UN No: UN1426; Haz Class: 4.3; Packing Group: I Powder; Linear Formula: NaBH4; MW: 37.83 Melting Point: 300° (decomposes) Density (g/mL): 1.074 Air Sensitive, Moisture Sensitive

  • RTECS: ED3325000
  • Air Sensitive, Moisture Sensitive
  • Powder


Formula: NaBH₄
MW: 37.83 g/mol
Melting Pt: >300 °C (dec.)
Density: 1.074 g/mL
Flash Pt: 69
MDL Number: MFCD00003518
CAS Number: 16940-66-2
EINECS: 241-004-4
UN: 1426
ADR: 4.3 I
Merck Index: 13,08664
Order Now
Specification Test Results
Assay 98%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty