Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Cyclopentene ≥98%

Supplier: BEANTOWN CHEMICAL - BTC

217470-100ML 217470-25ML
BT217470-100MLEA 114.78 USD InStock
BT217470-100ML BT217470-25ML
Cyclopentene ≥98%
Cyclopentene
CAS: 142-29-0; EC No: 205-532-9; MDL No: MFCD00001394; RTECS: GY5950000 UN No: UN2246; Haz Class: 3; Packing Group: II Liquid; Molecular Formula: C5H8; MW: 68.12 Melting Point: -135°; Boiling Point: 44-46°; Flash point: -34°C (-29°F) Density (g/mL): 0.771; Refractive Index: 1.421 Air Sensitive

  • RTECS: GY5950000
  • Refractive index: 1.421
  • Air Sensitive
  • Liquid


Formula: C₅H₈
MDL Number: MFCD00001394
CAS Number: 142-29-0
EINECS: 205-532-9
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty