Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Linear alkylbenzenesulfonic acid ≥97%

Supplier: BEANTOWN CHEMICAL - BTC

217850-1KG 217850-250G
BT217850-1KGEA 91.31 USD InStock
BT217850-1KG BT217850-250G
Linear alkylbenzenesulfonic acid ≥97%
Linear alkylbenzenesulfonic acid
CAS: 68584-22-5; EC No: 271-528-9; MDL No: MFCD00147445; RTECS: DB4290000 UN No: UN2586; Haz Class: 8; Packing Group: III Liquid

  • RTECS: DB4290000
  • Liquid


Formula: C₁₈H₃₀O₃S
MDL Number: MFCD00147445
CAS Number: 68584-22-5
EINECS: 271-528-9
ADR: 8,III
Order Now
Specification Test Results
Assay 97%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty