Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Bromocumene ≥97%

Supplier: BEANTOWN CHEMICAL - BTC

218025-25G 218025-5G
BT218025-25GEA 317 USD InStock
BT218025-25G BT218025-5G
2-Bromocumene ≥97%
2-Bromocumene
CAS: 7073-94-1; EC No: 230-370-0; MDL No: MFCD00051567 UN No: UN3082; Haz Class: 9; Packing Group: III Liquid; Molecular Formula: C9H11Br; MW: 199.10 Density (g/mL): 1.30; Refractive Index: 1.541

  • Refractive index: 1.541
  • Liquid


Formula: C₉H₁₁Br
MW: 199.09 g/mol
Density: 1.30 g/mL
MDL Number: MFCD00051567
CAS Number: 7073-94-1
EINECS: 230-370-0
UN: 3082
ADR: 9 III
Order Now
Specification Test Results
Assay 97%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty