Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Trifluoroacetic anhydride ≥99%

Supplier: BEANTOWN CHEMICAL - BTC

218030-100G 218030-25G 218030-500G
BT218030-100GEA 75.22 USD InStock
BT218030-100G BT218030-25G BT218030-500G
Trifluoroacetic anhydride ≥99%
Trifluoroacetic anhydride
CAS: 407-25-0; EC No: 206-982-9; MDL No: MFCD00000416; RTECS: AJ9800000 UN No: UN3265; Haz Class: 8; Packing Group: I Liquid; Linear Formula: (CF3CO)2O; Molecular Formula: C4F6O3; MW: 210.03 Melting Point: -65°; Boiling Point: 39-40° Density (g/mL): 1.511; Refractive Index: 1.269 Moisture Sensitive

  • RTECS: AJ9800000
  • Refractive index: 1.269
  • Moisture Sensitive
  • Liquid


Formula: C₄F₆O₃
MW: 210.03 g/mol
Boiling Pt: 39…40 °C
Melting Pt: –65 °C
Density: 1.511 g/mL
MDL Number: MFCD00000416
CAS Number: 407-25-0
EINECS: 206-982-9
UN: 3265
ADR: 8 I
Order Now
Specification Test Results
Assay 99%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty