Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,2-Difluoroethanol ≥95%

Supplier: BEANTOWN CHEMICAL - BTC

218450-1G 218450-5G
BT218450-1GEA 50 USD InStock
BT218450-1G BT218450-5G
2,2-Difluoroethanol ≥95%
2,2-Difluoroethanol
CAS: 359-13-7; MDL No: MFCD00042248 UN No: UN1987; Haz Class: 3; Packing Group: II Liquid; Linear Formula: HCF2CH2OH; MW: 82.05 Boiling Point: 95°; Flash point: 3°C (37°F) Density (g/mL): 1.296

  • Liquid


Formula: C₂H₄F₂O
MW: 82.05
Boiling Pt: 95 °C
Melting Pt: -28.2
Density: 1.296
Flash Pt: 3 °C (37 °F)
MDL Number: MFCD00042248
CAS Number: 359-13-7
UN: 1987
ADR: 3 II
Order Now
Specification Test Results
Assay 95%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty