Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

O,S-Diethyl methylphosphonothionate ≥97%

Supplier: BEANTOWN CHEMICAL - BTC

218645-2G 218645-500MG
BT218645-2GEA 1521 USD InStock
BT218645-2G BT218645-500MG
O,S-Diethyl methylphosphonothionate ≥97%
O,S-Diethyl methylphosphonothionate
CAS: 2511-10-6; MDL No: MFCD01705977; RTECS: TB1085000 UN No: UN3278; Haz Class: 6.1; Packing Group: I Liquid; Molecular Formula: C5H13O2PS; MW: 168.19 Boiling Point: 95-97°/10 mmHg Density (g/mL): 1.094; Refractive Index: 1.474

  • RTECS: TB1085000
  • Refractive index: 1.474
  • Liquid


Formula: C₅H₁₃O₂PS
MW: 168.19 g/mol
Boiling Pt: 95…97 °C/10 mmHg
Density: 1.094 g/mL
MDL Number: MFCD01705977
CAS Number: 2511-10-6
UN: 3278
ADR: 6.1 I
Order Now
Specification Test Results
Assay 97%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty