Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Benzyl(dimethyl)amine ≥98%

Supplier: BEANTOWN CHEMICAL - BTC

218785-1L 218785-250ML
BT218785-1LEA 89 USD InStock
BT218785-1L BT218785-250ML
Benzyl(dimethyl)amine ≥98%
Benzyl(dimethyl)amine
CAS: 103-83-3; EC No: 203-149-1; MDL No: MFCD00008329; RTECS: DP4500000 UN No: UN2619; Haz Class: 8 (3); Packing Group: II Liquid; Linear Formula: C6H5CH2N(CH3)2; Molecular Formula: C9H13N; MW: 135.21 Melting Point: -75°; Boiling Point: 183-184°; Flash point: 53°C (127°F) Density (g/mL): 0.9; Refractive Index: 1.501 Air Sensitive

  • RTECS: DP4500000
  • Refractive index: 1.501
  • Air Sensitive
  • Liquid


Formula: C₆H₅CH₂N(CH₃)₂
Boiling Pt: 183…184 °C
Melting Pt: –75 °C
Density: 0.9 g/mL
Flash Pt: 53 °C (127 °F)
MDL Number: MFCD00008329
CAS Number: 103-83-3
EINECS: 203-149-1
UN: 2619
ADR: 8,II
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=90635bbe74e3372ed4bfbd8d750b25afd50f77a5&vid=90635bbe74e3372ed4bfbd8d750b25afd50f77a5&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Assay 98%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty