Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Silver(I) oxide ≥99%

Supplier: BEANTOWN CHEMICAL - BTC

218890-100G 218890-25G 218890-500G
BT218890-100GEA 402.9 USD InStock
BT218890-100G BT218890-25G BT218890-500G
Silver(I) oxide ≥99%
Silver(I) oxide
CAS: 20667-12-3; EC No: 243-957-1; MDL No: MFCD00003404; RTECS: VW4900000 UN No: UN1479; Haz Class: 5.1; Packing Group: II Powder; Molecular Formula: Ag2O; MW: 231.74 Melting Point: 230° (decomposes) Density (g/mL): 7.143 Light Sensitive

  • RTECS: VW4900000
  • Light Sensitive


Formula: Ag₂O
Melting Pt: 230 °C (dec.)
Density: 7.143 g/mL
MDL Number: MFCD00003404
CAS Number: 20667-12-3
UN: 1479
ADR: 5.1,II
Order Now
Specification Test Results
Assay 99%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty