218975-100G
218975-25G
218975-500G
BT218975-100GEA
88
USD
InStock
BT218975-100G
BT218975-25G
BT218975-500G
1-(2-Methoxyphenyl)piperazine hydrochloride ≥97%
1-(2-Methoxyphenyl)piperazine hydrochloride
CAS: 5464-78-8; EC No: 226-762-6; MDL No: MFCD00012762
UN No: UN2811; Haz Class: 6.1; Packing Group: III
Solid; Molecular Formula: C11H16N2O·HCl ; MW: 228.72
Melting Point: 217-219°
Hygroscopic