219035-1G
219035-250MG
219035-5G
BT219035-1GEA
89
USD
InStock
BT219035-1G
BT219035-250MG
BT219035-5G
3-Trimethylsilylpropynal ≥97%
3-Trimethylsilylpropynal
CAS: 2975-46-4; MDL No: MFCD01103709
UN No: UN1993; Haz Class: 3; Packing Group: II
Liquid; Linear Formula: (CH3)3SiC≡CCHO ; Molecular Formula: C6H10OSi; MW: 126.23
Density (g/mL): 0.854; Refractive Index: 1.444
Air Sensitive, Moisture Sensitive
- Refractive index: 1.444
- Air Sensitive, Moisture Sensitive
- Liquid