Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Bromothiophene ≥98%

Supplier: BEANTOWN CHEMICAL - BTC

219195-1KG 219195-250G 219195-50G
BT219195-1KGEA 455 USD InStock
BT219195-1KG BT219195-250G BT219195-50G
2-Bromothiophene ≥98%
2-Bromothiophene
CAS: 1003-09-4; EC No: 213-699-4; MDL No: MFCD00005417 UN No: UN2929; Haz Class: 6.1 (3); Packing Group: II Liquid; Molecular Formula: C4H3BrS ; MW: 163.04 Boiling Point: 149-151°; Flash point: 52°C (126°F) Density (g/mL): 1.684; Refractive Index: 1.586 Light Sensitive

  • Refractive index: 1.586
  • Light Sensitive
  • Liquid


Formula: C₄H₃BrS
MW: 163.04 g/mol
Boiling Pt: 149
Melting Pt: -10
Density: 1.684 g/mL
Flash Pt: 52 °C (126 °F)
MDL Number: MFCD00005417
CAS Number: 1003-09-4
EINECS: 213-699-4
UN: 2929
ADR: 6.1 II
Order Now
Specification Test Results
Assay 98%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty