Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Dicyclohexylcarbodiimide ≥99%

Supplier: BEANTOWN CHEMICAL - BTC

219300-100G 219300-25G 219300-500G
BT219300-100GEA 49.22 USD InStock
BT219300-100G BT219300-25G BT219300-500G
Dicyclohexylcarbodiimide ≥99%
Dicyclohexylcarbodiimide
CAS: 538-75-0; EC No: 208-704-1; MDL No: MFCD00011659; RTECS: FF2160000 UN No: UN2811; Haz Class: 6.1; Packing Group: III Crystalline/Powder; Linear Formula: C6H11N=C=NC6H11; Molecular Formula: C13H22N2; MW: 206.33 Melting Point: 33-36°; Boiling Point: 122-124°/6 mmHg; Flash point: 110°C (230°F) Moisture Sensitive

  • RTECS: FF2160000
  • Moisture Sensitive
  • Crystalline/Powder


Formula: C₁₃H₂₂N₂
MW: 206.33 g/mol
Boiling Pt: 122…124 °C/6 mmHg
Melting Pt: 33…36 °C
Flash Pt: >110 °C (>230 °F)
MDL Number: MFCD00011659
CAS Number: 538-75-0
EINECS: 208-704-1
UN: 2811
ADR: 6.1 III
Merck Index: 13,03123
Order Now
Specification Test Results
Assay 99%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty