Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ammonium fluoride ≥98.0% ACS

Supplier: BEANTOWN CHEMICAL - BTC

219560-1KG 219560-250G
BT219560-1KGEA 217.38 USD InStock
BT219560-1KG BT219560-250G
Ammonium fluoride ≥98.0% ACS
Ammonium fluoride
CAS: 12125-01-8; EC No: 235-185-9; MDL No: MFCD00011423; RTECS: BQ6300000 UN No: UN2505; Haz Class: 6.1; Packing Group: III Crystalline; Linear Formula: NH4F; MW: 37.04 Hygroscopic

  • RTECS: BQ6300000
  • Hygroscopic
  • Crystalline


Formula: NH₄F
MW: 37.04 g/mol
Melting Pt: 160
MDL Number: MFCD00011423
CAS Number: 12125-01-8
EINECS: 235-185-9
UN: 2505
ADR: 6.1 III
Merck Index: 13,00524
Order Now
Specification Test Results
Assay 98.0% min.
Insoluble matter ≤0.005%
Residue after ignition ≤0.01%
Chloride (Cl) ≤0.001%
Sulfate (SO4) ≤0.005%
Heavy metals (as Pb) ≤5ppm
Iron (Fe) ≤5ppm

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty