Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Thallium(III) trifluoroacetate ≥95%

Supplier: BEANTOWN CHEMICAL - BTC

219730-1G 219730-5G
BT219730-1GEA 17 USD InStock
BT219730-1G BT219730-5G
Thallium(III) trifluoroacetate ≥95%
Thallium(III) trifluoroacetate
CAS: 23586-53-0; EC No: 245-761-1; MDL No: MFCD00000414 UN No: UN1707; Haz Class: 6.1; Packing Group: II Solid; Linear Formula: Tl(OOCCF3)3; Molecular Formula: C6F9O6Tl; MW: 543.42 Melting Point: 213° (decomposes) Air Sensitive , Hygroscopic

  • Air Sensitive, Hygroscopic
  • Solid


Formula: C₆F₉O₆Tl
MW: 543.43 g/mol
Boiling Pt: 160
Melting Pt: 120.5 (dec.)
MDL Number: MFCD00000414
CAS Number: 23586-53-0
EINECS: 245-761-1
UN: 1707
ADR: 6.1 II
Order Now
Specification Test Results
Assay 95%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty