220035-100G
220035-25G
220035-500G
BT220035-100GEA
102.02
USD
InStock
BT220035-100G
BT220035-25G
BT220035-500G
Aminomethanesulfonic acid ≥97%
Aminomethanesulphonic acid
CAS: 13881-91-9; EC No: 237-649-6; MDL No: MFCD00008124
UN No: UN2585; Haz Class: 8; Packing Group: III
Powder; Linear Formula: NH2CH2SO3H; Molecular Formula: CH5NO3S; MW: 111.12
Melting Point: 184° (decomposes)